accessibility menu, dialog, popup

UserName

CH3OOHNHCl

CAS RN 65365-28-8

IUPAC Name: methyl pyrrolidine-2-carboxylate hydrochloride
Synonyms: methyl pyrrolidine-2-carboxylate hydrochloride
Molecular Weight (g/mol): 165.62
Molecular Formula: C6H12ClNO2
InChi Key: HQEIPVHJHZTMDP-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C1CCCN1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)