missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Proline Methyl Ester Hydrochloride 96.0+%, TCI America™
$54.06 - $150.98
Chemical Identifiers
| CAS | 65365-28-8 |
|---|---|
| Molecular Formula | C6H12ClNO2 |
| Molecular Weight (g/mol) | 165.617 |
| MDL Number | MFCD00083685 |
| InChI Key | HQEIPVHJHZTMDP-NUBCRITNSA-N |
| Synonym | d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride |
| PubChem CID | 44629857 |
| IUPAC Name | methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
| SMILES | COC(=O)C1CCCN1.Cl |
Chemical Identifiers
| 65365-28-8 | |
| 165.617 | |
| HQEIPVHJHZTMDP-NUBCRITNSA-N | |
| 44629857 | |
| COC(=O)C1CCCN1.Cl |
| C6H12ClNO2 | |
| MFCD00083685 | |
| d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride | |
| methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride |
Specifications
| 65365-28-8 | |
| White | |
| MFCD00083685 | |
| d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride | |
| COC(=O)C1CCCN1.Cl | |
| 165.617 | |
| 165.62 | |
| Crystalline Powder |
| 73°C | |
| C6H12ClNO2 | |
| 1 g | |
| HQEIPVHJHZTMDP-NUBCRITNSA-N | |
| methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride | |
| 44629857 | |
| ≥96.0% (N,T) | |
| D-Proline Methyl Ester Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator