missing translation for 'onlineSavingsMsg'
Learn More

D-Proline Methyl Ester Hydrochloride 96.0+%, TCI America™

Catalog No. P17301G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17301G 1 g
P17305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17301G Supplier TCI America Supplier No. P17301G

Chemical Identifiers

CAS 65365-28-8
Molecular Formula C6H12ClNO2
Molecular Weight (g/mol) 165.617
MDL Number MFCD00083685
InChI Key HQEIPVHJHZTMDP-NUBCRITNSA-N
Synonym d-proline methyl ester hydrochloride, h-d-pro-ome.hcl, r-methyl pyrrolidine-2-carboxylate hydrochloride, h-d-pro-ome hcl, unii-c15epu1m46, d-proline, methyl ester, hydrochloride, methyl d-prolinate hydrochloride, r-proline methyl ester hydrochloride, methyl r-pyrrolidine-2-carboxylate hydrochloride, methyl 2r-pyrrolidine-2-carboxylate hydrochloride
PubChem CID 44629857
IUPAC Name methyl (2R)-pyrrolidine-2-carboxylate;hydrochloride
SMILES COC(=O)C1CCCN1.Cl

Specifications

Melting Point 73°C
Color White
Quantity 1 g
Formula Weight 165.62
Percent Purity ≥96.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Proline Methyl Ester Hydrochloride
TSCA No
Recommended Storage Refrigerator