missing translation for 'onlineSavingsMsg'
Learn More

D-Pinitol, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012328
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012328 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012328 Supplier Merck Emd Millipore Supplier No. 7494825MG
Only null left

Specifications

CAS 10284-63-6
Linear Formula C7H14O6
MDL Number MFCD00216659
Melting Point 179°C to 185°C
Synonym 3-O-Methyl-D-chiro-inositol
Type Analytical Standard
Quantity 25 mg
Formula Weight 194.18
Percent Purity ≥98.0% (GC)
Grade Analytical Standard