missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Phenylalaninol 98.0+%, TCI America™
Supplier: TCI America P128925G
Specifications
| D-Phenylalaninol | |
| 93°C | |
| C9H13NO | |
| 25 g | |
| d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol | |
| C1=CC=C(C=C1)CC(CO)N | |
| 151.209 | |
| 151.21 | |
| Crystalline Powder |
| 5267-64-1 | |
| 122°C | |
| MFCD00064298 | |
| 3259 | |
| STVVMTBJNDTZBF-SECBINFHSA-N | |
| (2R)-2-amino-3-phenylpropan-1-ol | |
| 853475 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 5267-64-1 | |
| 151.209 | |
| STVVMTBJNDTZBF-SECBINFHSA-N | |
| 853475 | |
| C1=CC=C(C=C1)CC(CO)N |
| C9H13NO | |
| MFCD00064298 | |
| d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol | |
| (2R)-2-amino-3-phenylpropan-1-ol |
Safety and Handling
TSCA : No