missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Phenylalaninol 98.0+%, TCI America™
$134.21 - $406.37
Chemical Identifiers
| CAS | 5267-64-1 |
|---|---|
| Molecular Formula | C9H13NO |
| Molecular Weight (g/mol) | 151.209 |
| MDL Number | MFCD00064298 |
| InChI Key | STVVMTBJNDTZBF-SECBINFHSA-N |
| Synonym | d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol |
| PubChem CID | 853475 |
| IUPAC Name | (2R)-2-amino-3-phenylpropan-1-ol |
| SMILES | C1=CC=C(C=C1)CC(CO)N |
Chemical Identifiers
| 5267-64-1 | |
| 151.209 | |
| STVVMTBJNDTZBF-SECBINFHSA-N | |
| 853475 | |
| C1=CC=C(C=C1)CC(CO)N |
| C9H13NO | |
| MFCD00064298 | |
| d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol | |
| (2R)-2-amino-3-phenylpropan-1-ol |
Specifications
| 5267-64-1 | |
| 122°C | |
| MFCD00064298 | |
| 3259 | |
| STVVMTBJNDTZBF-SECBINFHSA-N | |
| (2R)-2-amino-3-phenylpropan-1-ol | |
| 853475 | |
| ≥98.0% (GC,T) | |
| D-Phenylalaninol |
| 93°C | |
| C9H13NO | |
| 5 g | |
| d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol | |
| C1=CC=C(C=C1)CC(CO)N | |
| 151.209 | |
| 151.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : No