missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalanine tert-butyl ester hydrochloride, 98%

Catalog No. p-7054892
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6332503 1 g
AAH6332506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6332503 Supplier Thermo Scientific Chemicals Supplier No. H6332503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3403-25-6 | C13H20ClNO2 | 257.758 g/mol

D-Phenylalanine tert-butyl ester hydrochloride is used as a fine chemical intermediate, agrochemical &pharmceutical intermediate. It has anti - static and anti- corrosion properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Phenylalanine tert-butyl ester hydrochloride is used as a fine chemical intermediate, agrochemical &pharmceutical intermediate. It has anti - static and anti- corrosion properties.

Solubility
Soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 3403-25-6
Molecular Formula C13H20ClNO2
Molecular Weight (g/mol) 257.758
MDL Number MFCD00153455
InChI Key FDMCEXDXULPJPG-RFVHGSKJSA-N
Synonym h-d-phe-otbu.hcl, r-tert-butyl 2-amino-3-phenylpropanoate hydrochloride, tert-butyl 2r-2-amino-3-phenylpropanoate hydrochloride, d-phenylalanine tert-butyl ester hydrochloride, d-phenylalanine t-butylester hydrochloride, d-phenylalanine, 1,1-dimethylethyl ester, hydrochloride, h-d-phe-otbu hydrochloride, tert-butyl d-phenylalaninate hydrochloride, h-d-phe-otbu hcl, h-d-phe-obut hcl
PubChem CID 15942423
IUPAC Name tert-butyl (2R)-2-amino-3-phenylpropanoate;hydrochloride
SMILES CC(C)(C)OC(=O)C(CC1=CC=CC=C1)N.Cl

Specifications

Solubility Information Soluble in water.
Quantity 1 g
Formula Weight 257.76
Percent Purity 98%
Physical Form Solid
Chemical Name or Material D-Phenylalanine tert-butyl ester hydrochloride
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only