missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Phenylalanine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™
Supplier: TCI America P172225G
Specifications
| D-Phenylalanine Benzyl Ester p-Toluenesulfonate | |
| 169°C | |
| C23H25NO5S | |
| 25 g | |
| ZLZGBBIPWXUQST-UHFFFAOYNA-N | |
| 4-methylbenzene-1-sulfonic acid; benzyl 2-amino-3-phenylpropanoate | |
| 44629866 | |
| ≥98.0% (HPLC,T) |
| 28607-46-7 | |
| White | |
| MFCD00066129 | |
| h-d-phe-obzl tos, d-phenylalanine benzyl ester p-toluenesulfonate, h-d-phe-obzl.tosoh, d-phenylalanine benzyl ester tosylate, r-benzyl 2-amino-3-phenylpropanoate 4-methylbenzenesulfonate, benzyl 2r-2-amino-3-phenylpropanoate; para-toluene sulfonate, d-phe-obzl, h-d-phe-obzl.tos, h-d-phe-obzl?os, h-d-phe-obzl. tos | |
| CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 | |
| 427.52 | |
| 427.52 | |
| Crystalline Powder |
Chemical Identifiers
| 28607-46-7 | |
| 427.52 | |
| ZLZGBBIPWXUQST-UHFFFAOYNA-N | |
| 44629866 | |
| CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 |
| C23H25NO5S | |
| MFCD00066129 | |
| h-d-phe-obzl tos, d-phenylalanine benzyl ester p-toluenesulfonate, h-d-phe-obzl.tosoh, d-phenylalanine benzyl ester tosylate, r-benzyl 2-amino-3-phenylpropanoate 4-methylbenzenesulfonate, benzyl 2r-2-amino-3-phenylpropanoate; para-toluene sulfonate, d-phe-obzl, h-d-phe-obzl.tos, h-d-phe-obzl?os, h-d-phe-obzl. tos | |
| 4-methylbenzene-1-sulfonic acid; benzyl 2-amino-3-phenylpropanoate |
Safety and Handling
TSCA : No