missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalanine Benzyl Ester p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. p-7135225
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17225G 5 g
P172225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17225G Supplier TCI America Supplier No. P17225G

Chemical Identifiers

CAS 28607-46-7
Molecular Formula C23H25NO5S
Molecular Weight (g/mol) 427.52
MDL Number MFCD00066129
InChI Key ZLZGBBIPWXUQST-UHFFFAOYNA-N
Synonym h-d-phe-obzl tos, d-phenylalanine benzyl ester p-toluenesulfonate, h-d-phe-obzl.tosoh, d-phenylalanine benzyl ester tosylate, r-benzyl 2-amino-3-phenylpropanoate 4-methylbenzenesulfonate, benzyl 2r-2-amino-3-phenylpropanoate; para-toluene sulfonate, d-phe-obzl, h-d-phe-obzl.tos, h-d-phe-obzl?os, h-d-phe-obzl. tos
PubChem CID 44629866
IUPAC Name 4-methylbenzene-1-sulfonic acid; benzyl 2-amino-3-phenylpropanoate
SMILES CC1=CC=C(C=C1)S(O)(=O)=O.NC(CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 169°C
Color White
Quantity 5 g
Formula Weight 427.52
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-Phenylalanine Benzyl Ester p-Toluenesulfonate