missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Pantothenic Acid Sodium Salt, Spectrum™ Chemical
P1468, 867-81-2, C9H16NO5Na
$394.94 - $11925.39
Chemical Identifiers
| CAS | 867-81-2 |
|---|---|
| Molecular Formula | C9H16NNaO5 |
| Molecular Weight (g/mol) | 241.22 |
| MDL Number | MFCD00002767 |
| InChI Key | ROMBWECPDOCQMZ-UHFFFAOYNA-M |
| IUPAC Name | sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate |
| SMILES | [Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602346
|
Spectrum Chemical Mfg Cor
P146825GM |
25 g | Amber Glass Bottle |
Each for $394.94
|
|
||||
|
18602347
|
Spectrum Chemical Mfg Cor
P1468100GM |
100 g | Amber Glass Bottle |
Each for $1,213.18
|
|
||||
|
18602348
|
Spectrum Chemical Mfg Cor
P14681KGBL |
1 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical D-Pantothenic Acid Sodium Salt is also known as pantothenate and is soluble in water. It is considered an essential nutrient for several types of animals. It is also an ingredient in some skin or hair products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 867-81-2 | |
| 241.22 | |
| ROMBWECPDOCQMZ-UHFFFAOYNA-M | |
| [Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O |
| C9H16NNaO5 | |
| MFCD00002767 | |
| sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate |
Specifications
| 867-81-2 | |
| C9H16NNaO5 | |
| MFCD00002767 | |
| [Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O | |
| 25 g | |
| Technical |
| 100% | |
| Amber Glass Bottle | |
| ROMBWECPDOCQMZ-UHFFFAOYNA-M | |
| sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate | |
| 241.22 |