missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Pantothenic Acid Sodium Salt, Spectrum™ Chemical
P1468, 867-81-2, C9H16NO5Na
Supplier: Spectrum Chemical Mfg Cor P1468100GM
Description
Spectrum™ Chemical D-Pantothenic Acid Sodium Salt is also known as pantothenate and is soluble in water. It is considered an essential nutrient for several types of animals. It is also an ingredient in some skin or hair products. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 867-81-2 | |
| C9H16NNaO5 | |
| MFCD00002767 | |
| [Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O | |
| 100 g | |
| Technical |
| 100% | |
| Amber Glass Bottle | |
| ROMBWECPDOCQMZ-UHFFFAOYNA-M | |
| sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate | |
| 241.22 |
Chemical Identifiers
| 867-81-2 | |
| 241.22 | |
| ROMBWECPDOCQMZ-UHFFFAOYNA-M | |
| [Na+].CC(C)(O)CC(O)C(=O)NCCC([O-])=O |
| C9H16NNaO5 | |
| MFCD00002767 | |
| sodium 3-(2,4-dihydroxy-4-methylpentanamido)propanoate |