missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Pantothenic acid calcium salt, Cell culture reagent grade, MP Biomedicals™
$129.79 - $129.79
Chemical Identifiers
| CAS | 2219-30-9 |
|---|---|
| Molecular Formula | C5H12ClNO2S |
| Molecular Weight (g/mol) | 185.666 |
| InChI Key | CZDHUFYOXKHLME-DFWYDOINSA-N |
| Synonym | d-penicillamine hydrochloride, penicillamine hydrochloride, distamine, unii-5n9jk529zj, usaf el-23, usaf a-1705, d---penicillamine hydrochloride, d-valine, 3-mercapto-, hydrochloride, valine, 3-mercapto-, hydrochloride, d |
| PubChem CID | 92173 |
| IUPAC Name | (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid;hydrochloride |
| SMILES | CC(C)(C(C(=O)O)N)S.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19472180
|
MP Biomedicals Inc
0219472180 |
100 g |
Each for $129.79
|
|
|||||
Chemical Identifiers
| 2219-30-9 | |
| 185.666 | |
| d-penicillamine hydrochloride, penicillamine hydrochloride, distamine, unii-5n9jk529zj, usaf el-23, usaf a-1705, d---penicillamine hydrochloride, d-valine, 3-mercapto-, hydrochloride, valine, 3-mercapto-, hydrochloride, d | |
| (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid;hydrochloride |
| C5H12ClNO2S | |
| CZDHUFYOXKHLME-DFWYDOINSA-N | |
| 92173 | |
| CC(C)(C(C(=O)O)N)S.Cl |
Specifications
| 2219-30-9 | |
| C5H12ClNO2S | |
| CZDHUFYOXKHLME-DFWYDOINSA-N | |
| (2S)-2-amino-3-methyl-3-sulfanylbutanoic acid;hydrochloride | |
| 92173 | |
| Cell Culture Reagent | |
| D-Pantothenic acid |
| 100 g | |
| d-penicillamine hydrochloride, penicillamine hydrochloride, distamine, unii-5n9jk529zj, usaf el-23, usaf a-1705, d---penicillamine hydrochloride, d-valine, 3-mercapto-, hydrochloride, valine, 3-mercapto-, hydrochloride, d | |
| CC(C)(C(C(=O)O)N)S.Cl | |
| 185.666 | |
| 238.3 | |
| 0.5g/cm3 |
Safety and Handling
EINECSNumber : 205-278-9
Recommended Storage : Store at 2° to 8°C