missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Norgestrel, ≥99%, MilliporeSigma™ Supelco™

Catalog No. 111012243
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012243 100 mg
111012242 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012243 Supplier Merck Emd Millipore Supplier No. N2260100MG

D(−)-Norgestrel is a hormonal medication and a contraceptive used by women.

Specifications

Packaging Poly Bottle
CAS 797-63-7
Physical Form Neat
Linear Formula C21H28O2
MDL Number MFCD00199013
Synonym 13 beta-Ethyl-17 alpha-ethynyl-17 beta-hydroxygon-4-en-3-one; 17 alpha-Ethynyl-18-homo-19-nor-testosterone; 18,19-Dinor-13 beta-ethyl-17 beta-hydroxy-4-pregnen-20-yn-3-one; 18,19-Dinor-4-pregnen-20-yn-3-one; Levonorgestrel
Type Analytical Standard
Quantity 100 mg
Formula Weight 312.45
Percent Purity ≥99%
Grade Analytical Standard
Show More Show Less