missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Methionine 99.0+%, TCI America™
$136.70 - $467.07
Chemical Identifiers
| CAS | 348-67-4 |
|---|---|
| Molecular Formula | C5H11NO2S |
| Molecular Weight (g/mol) | 149.21 |
| MDL Number | MFCD00002622 |
| InChI Key | FFEARJCKVFRZRR-SCSAIBSYSA-N |
| Synonym | d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid |
| PubChem CID | 84815 |
| ChEBI | CHEBI:16867 |
| IUPAC Name | (2R)-2-amino-4-(methylsulfanyl)butanoic acid |
| SMILES | CSCC[C@@H](N)C(O)=O |
Chemical Identifiers
| C5H11NO2S | |
| MFCD00002622 | |
| d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
| CHEBI:16867 | |
| CSCC[C@@H](N)C(O)=O |
Specifications
| 348-67-4 | |
| C5H11NO2S | |
| d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid | |
| CSCC[C@@H](N)C(O)=O | |
| 149.21 | |
| CHEBI:16867 | |
| ≥99.0% (T) | |
| D-Methionine |
| White | |
| MFCD00002622 | |
| FFEARJCKVFRZRR-SCSAIBSYSA-N | |
| (2R)-2-amino-4-(methylsulfanyl)butanoic acid | |
| 84815 | |
| 149.21 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1254
RTECSNumber : PD0455000
TSCA : Yes