missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Melezitose Hydrate 99.0+%, TCI America™
$177.08 - $551.76
Chemical Identifiers
| CAS | 207511-10-2 |
|---|---|
| Molecular Formula | C18H32O16 |
| Molecular Weight (g/mol) | 504.44 |
| MDL Number | MFCD00006627 |
| InChI Key | QWIZNVHXZXRPDR-IXUUBQBONA-N |
| Synonym | d-+-melezitose hydrate |
| PubChem CID | 126963632 |
| IUPAC Name | (3R,4S,5S,6R)-2-{[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-3-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
| SMILES | OC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O |
Chemical Identifiers
| 207511-10-2 | |
| 504.44 | |
| QWIZNVHXZXRPDR-IXUUBQBONA-N | |
| 126963632 | |
| OC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O |
| C18H32O16 | |
| MFCD00006627 | |
| d-+-melezitose hydrate | |
| (3R,4S,5S,6R)-2-{[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-3-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
Specifications
| 207511-10-2 | |
| White | |
| MFCD00006627 | |
| d-+-melezitose hydrate | |
| OC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O | |
| 504.44 | |
| 504.44 | |
| Crystalline Powder |
| 153°C | |
| C18H32O16 | |
| 5 g | |
| QWIZNVHXZXRPDR-IXUUBQBONA-N | |
| (3R,4S,5S,6R)-2-{[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-3-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 126963632 | |
| ≥99.0% (GC) | |
| D-(+)-Melezitose Hydrate |
Safety and Handling
TSCA : Yes