missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Melezitose Hydrate 99.0+%, TCI America™
Supplier: TCI America M004925G
Specifications
| D-(+)-Melezitose Hydrate | |
| 153°C | |
| C18H32O16 | |
| 25 g | |
| QWIZNVHXZXRPDR-IXUUBQBONA-N | |
| (3R,4S,5S,6R)-2-{[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-3-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 126963632 | |
| ≥99.0% (GC) |
| 207511-10-2 | |
| White | |
| MFCD00006627 | |
| d-+-melezitose hydrate | |
| OC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O | |
| 504.44 | |
| 504.44 | |
| Crystalline Powder |
Chemical Identifiers
| 207511-10-2 | |
| 504.44 | |
| QWIZNVHXZXRPDR-IXUUBQBONA-N | |
| 126963632 | |
| OC[C@H]1O[C@@](CO)(OC2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H]1O |
| C18H32O16 | |
| MFCD00006627 | |
| d-+-melezitose hydrate | |
| (3R,4S,5S,6R)-2-{[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-3-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol |
Safety and Handling
TSCA : Yes