missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Mannose 98.0+%, TCI America™

Catalog No. M0045500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0045500G 500 g
M004525G 25 g
M0045100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0045500G Supplier TCI America Supplier No. M0045500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6H12O6
MDL Number MFCD00064122
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Color White
Quantity 500 g
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Mannose
EINECSNumber (8)-0685
TSCA Yes