Learn More
Thermo Scientific Chemicals D-(+)-Mannose, 99%
Description
D-mannose acts as an alternative energy source utilized by brain. It is used in the study of assessing the synthesis of amphiphilic glycopolymers and investigating early detection of bronchiolitis obliterans.It may be useful in the treatment of urinary tract infections (UTIs) and carbohydrate-deficient glycoprotein syndrome type 1b. It is also useful in the formation of glycan structure and glycosylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-mannose acts as an alternative energy source utilized by brain. It is used in the study of assessing the synthesis of amphiphilic glycopolymers and investigating early detection of bronchiolitis obliterans.It may be useful in the treatment of urinary tract infections (UTIs) and carbohydrate-deficient glycoprotein syndrome type 1b. It is also useful in the formation of glycan structure and glycosylation.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 127°C to 133°C |
| Density | 1.54 g/mL |
| Assay Percent Range | 99% |
| Molecular Formula | C6H12O6 |
| Quantity | 25 g |
| Beilstein | 1564373 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| Formula Weight | 180.16 |
| Chemical Name or Material | D-(+)-Mannose |
Safety and Handling
| EINECSNumber | 222-392-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only