missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Mannose, 99+%

CAS: 3458-28-4 | C6H12O6 | 180.16 g/mol

Supplier:  Thermo Scientific Chemicals 150600025

 View more versions of this product

Catalog No. AC150600025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D(+)-Mannose
133.0°C
0.45% max.
98% min. (ELSD) (HPLC)
Plastic drum
2.5 kg
155812
+ 14.00
Solubility in water: 2480g/L (17°C)
180.16
Crystals or Powder
3458-28-4
White
Authentic
Odorless
C6H12O6
31284
+ 14.00 (20.00°C 589nm,c=10,H2O)
2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers
180.16
99+%

Chemical Identifiers

3458-28-4
C6H12O6
180.16
2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

EINECSNumber : 222-392-4

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only