missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Mannose, 99+%

Catalog No. AC150601000
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150601000 100 g Plastic bottle
AC150600250 25 g Plastic bottle
AC150600010 1 kg Plastic bottle
AC150600025 2.5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC150601000 Supplier Thermo Scientific Chemicals Supplier No. 150601000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3458-28-4 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Melting Point 133.0°C
Color White
Loss on Drying 0.45% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Odor Odorless
Packaging Plastic bottle
Quantity 100 g
Beilstein 31284
Merck Index 155812
Specific Rotation Condition + 14.00 (20.00°C 589nm,c=10,H2O)
Specific Rotation + 14.00
Solubility Information Solubility in water: 2480g/L (17°C)
Formula Weight 180.16
Percent Purity 99+%
Physical Form Crystals or Powder
Chemical Name or Material D(+)-Mannose
Show More Show Less
EINECSNumber 222-392-4

RUO – Research Use Only