missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol, MP Biomedicals™

Catalog No. MP218379090
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP218379090 500 g
MP218379091 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP218379090 Supplier MP Biomedicals Inc Supplier No. 0218379090

D-Mannitol, ACS

  • Used in sweetness inhibition studies.
  • A sugar alcohol sweet tastant.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym Mannite
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 167°C to 170°C (literature)
Quantity 500 g
Grade ACS
storageNote1 Store at Room Temperature(15-30°C).