missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol, ACS Reagent, Reagents

Catalog No. 01337903
Change view
Click to view available options
Quantity:
1 kg
2 kg
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337903 1 kg
01337902 2 kg
01337901 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337903 Supplier Labchem Inc Supplier No. C12431001KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D-Mannitol, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym Mannitol
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Color White
Boiling Point 290°C
Packaging Poly Bottle
Quantity 1 kg
Specific Gravity 1.52
Grade ACS
Physical Form Solid
Chemical Name or Material D-Mannitol, ACS Reagent