missing translation for 'onlineSavingsMsg'
Learn More

D-mannitol, 99%, MP Biomedicals™

Catalog No. ICN10224801
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10224801 1 kg
ICN10224805 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10224801 Supplier MP Biomedicals Inc Supplier No. 0210224801
Only null left

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Density 1.52g/mL
Quantity 1 kg
IUPAC Name hexane-1,2,3,4,5,6-hexol
Formula Weight 182.2
Percent Purity 0.984
Chemical Name or Material D-Nannitol