missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Mannitol, 98+%

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol

Supplier:  Thermo Scientific Chemicals 125340025

Quantity 2.5 kg
Packaging Plastic bottle

 View more versions of this product

Catalog No. AC125340025


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

69-65-8
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
6251
OCC(O)C(O)C(O)C(O)CO
C6H14O6
MFCD00064287
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
CHEBI:16899

Specifications

D-Mannitol
165.0°C to 170.0°C
0.3% max. (105°C, 4 hrs)
98% min. (ELSD) (HPLC)
C6H14O6
MFCD00064287
15, 5810
+23° to +25° (20°C, 589nm) (PHEUR)
FBPFZTCFMRRESA-UHFFFAOYNA-N
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
6251
182.17
Crystalline Powder
69-65-8
White
Authentic
Plastic bottle
2.5 kg
01, IV, 2841
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents
OCC(O)C(O)C(O)C(O)CO
182.17
CHEBI:16899
98+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-711-8

SDS
Documents

Documents

Promotions

Promotions