missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Mannitol, 98+%

Catalog No. AC125340010
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125340010 1 kg Plastic bottle
AC125341000 100 g Plastic bottle
AC125345000 500 g Plastic bottle
AC125340025 2.5 kg Plastic bottle
AC125340050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC125340010 Supplier Thermo Scientific Chemicals Supplier No. 125340010

missing translation for 'validMainframeMsg'

CAS: 69-65-8 | C6H14O6 | 182.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 165.0°C to 170.0°C
Color White
Loss on Drying 0.3% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 01, IV, 2841
Merck Index 15, 5810
Specific Rotation +23° to +25° (20°C, 589nm) (PHEUR)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
Formula Weight 182.17
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material D-Mannitol
Show More Show Less
EINECSNumber 200-711-8

RUO – Research Use Only