missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Maltose Monohydrate, ≥92% (Dry Basis), Ultrapure

Catalog No. p-7082847
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ18725A1 1 kg Plastic bottle
AAJ18725A7 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ18725A1 Supplier Thermo Fisher Scientific Supplier No. J18725A1
Only null left

D-(+)-Maltose Monohydrate, >-92% (Dry Basis), Ultrapure, Affymetrix/USB, Quantity: 5kg, Packaging: Plastic bottle, Grade: Ultrapure, Melting Point: ∼120 deg.C (decomposition), Percent Purity: >-92%, Assay Percent Notes: Dry basis, Beilstein: 5784659, CAS: 6363-53-7, EINECS Number: 200-716-5 | CAS: 6363-53-7 | C12H22O11·H2O | 360.31 g/mol

D-(+)-Maltose Monohydrate, >-92% (Dry Basis), Ultrapure, Affymetrix/USB, Quantity: 5kg, Packaging: Plastic bottle, Grade: Ultrapure, Melting Point: ∼120 deg.C (decomposition), Percent Purity: >-92%, Assay Percent Notes: Dry basis, Beilstein: 5784659, CAS: 6363-53-7, EINECS Number: 200-716-5

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H22O11·H2O
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
Synonym d-+-maltose monohydrate, unii-dm477ee40d, 2 r,3 r,4 r,5 r-2,3,5,6-tetrahydroxy-4-2 r,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

Melting Point ∼120°C (decomposition)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 5784659
Formula Weight 360.31
Percent Purity ≥92%
Grade Ultrapure
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material D-(+)-Maltose Monohydrate
EINECSNumber 200-716-5
RTECSNumber OO5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only