missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Maltose monohydrate, 92+%

Catalog No. AC329911000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC329911000 100 g Plastic bottle
AC329915000 500 g Plastic bottle
AC329910025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC329911000 Supplier Thermo Scientific Chemicals Supplier No. 329911000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6363-53-7 | C12H24O12 | 360.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
Synonym d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

Melting Point 130.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 92% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 31,386
Merck Index 14,5714
Specific Rotation Condition + 137.00 (20.00°C c=4,H2O,NH3)
Specific Rotation + 137.00
Solubility Information Solubility in water: 1080g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in ether
Formula Weight 360.29
Percent Purity 92+%
Physical Form Fine Crystalline Powder or Crystals
Chemical Name or Material D(+)-Maltose monohydrate, 90%
Show More Show Less

RUO – Research Use Only