missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-(+)-Malic Acid 98.0+%, TCI America™
Supplier: TCI America M002125G
Specifications
| D-(+)-Malic Acid | |
| 103°C | |
| C4H6O5 | |
| 25 g | |
| BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
| 2-hydroxybutanedioic acid | |
| 92824 | |
| 134.09 | |
| Crystalline Powder |
| 636-61-3 | |
| White | |
| MFCD00004245 | |
| d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate | |
| OC(CC(O)=O)C(O)=O | |
| 134.09 | |
| CHEBI:30796 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 636-61-3 | |
| 134.09 | |
| BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
| 92824 | |
| 2-hydroxybutanedioic acid |
| C4H6O5 | |
| MFCD00004245 | |
| d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate | |
| CHEBI:30796 | |
| OC(CC(O)=O)C(O)=O |
Safety and Handling
EINECSNumber : (2)-1442
RTECSNumber : ON7260000
TSCA : No