missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Malic Acid 98.0+%, TCI America™

Catalog No. p-7131931
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M00215G 5 g
M002125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M00215G Supplier TCI America Supplier No. M00215G
Only null left

Chemical Identifiers

CAS 636-61-3
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
MDL Number MFCD00004245
InChI Key BJEPYKJPYRNKOW-UHFFFAOYNA-N
Synonym d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate
PubChem CID 92824
ChEBI CHEBI:30796
IUPAC Name 2-hydroxybutanedioic acid
SMILES OC(CC(O)=O)C(O)=O

Specifications

Melting Point 103°C
Color White
Quantity 5 g
Formula Weight 134.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Malic Acid
EINECSNumber (2)-1442
RTECSNumber ON7260000
TSCA No