missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Lysine monohydrochloride, 98%

Catalog No. AAL0771003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0771003 1 g
AAL0771006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0771003 Supplier Thermo Scientific Chemicals Supplier No. L0771003

CAS: 7274-88-6 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7274-88-6
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012920
InChI Key BVHLGVCQOALMSV-XZNNNFJINA-N
Synonym d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d
PubChem CID 81691
ChEBI CHEBI:53558
IUPAC Name (2R)-2,6-diaminohexanoic acid;hydrochloride
SMILES Cl.NCCCC[C@@H](N)C(O)=O

Specifications

Melting Point ∽260°C (decomposition)
Beilstein 4356907
Quantity 1 g
Formula Weight 182.65
Optical Rotation −20° (c=5 in 5N HCl)
Percent Purity 98%
Chemical Name or Material D-Lysine monohydrochloride
EINECSNumber 230-691-6
RTECSNumber OL5632500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only