missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Lysine hydrochloride, 99+%

Catalog No. p-3776608
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC243080010 1 g Glass Bottle
AC243080100 10 g Glass Bottle
AC243080500 50 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC243080010 Supplier Thermo Scientific Chemicals Supplier No. 243080010

CAS: 7274-88-6 | C6H15ClN2O2 | 182.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7274-88-6
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012920
InChI Key BVHLGVCQOALMSV-XZNNNFJINA-N
Synonym d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d
PubChem CID 81691
ChEBI CHEBI:53558
SMILES Cl.NCCCC[C@@H](N)C(O)=O

Specifications

Arsenic (As) 1ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99+%
Loss on Drying 0.3% max. (105°C, 3 hrs)
Packaging Glass Bottle
Linear Formula H2N(CH2)4CH(NH2)COOH·HCl
Specific Rotation -20.5° to -21.5° (20°C, 589nm) (c=8, 6N HCl)
Fieser 01,959; 07,79; 01,977
Sulfated Ash 0.1% max.
IUPAC Name (2R)-2,6-diaminohexanoic acid hydrochloride
Specific Rotation Condition −21° (20°C c=8,6N HCl)
Quantity 1 g
Formula Weight 182.65
Percent Purity 99.0 to 101.0%
Physical Form Crystalline Powder
Chemical Name or Material D-Lysine hydrochloride
Show More Show Less

RUO – Research Use Only