missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Luciferin

Catalog No. p-154155
Change view
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC337050100 10 mg Glass bottle
AC337050500 50 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC337050100 Supplier Thermo Scientific Chemicals Supplier No. 337050100

CAS: 2591-17-5 | C11H8N2O3S2 | 280.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2591-17-5
Molecular Formula C11H8N2O3S2
Molecular Weight (g/mol) 280.3
MDL Number MFCD00042929
InChI Key IWJYWBVPCGUPLO-SSDOTTSWSA-N
Synonym luciferin, d-luciferin, s-2-6-hydroxybenzo d thiazol-2-yl-4,5-dihydrothiazole-4-carboxylic acid, luciferin firefly, s-2-6-hydroxy-2-benzothiazolyl-2-thiazoline-4-carboxylic acid, 4s-2-6-oxo-1,3-benzothiazol-2-ylidene-1,3-thiazolidine-4-carboxylic acid, 4-thiazolecarboxylicacid, 4,5-dihydro-2-6-hydroxy-2-benzothiazolyl-, 4s
PubChem CID 7023066
IUPAC Name (4S)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylic acid
SMILES C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)O

Specifications

Melting Point 200°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10 mg
Merck Index 15, 4117
Specific Rotation -32.0° to -36.0° (20°C, 589nm) (c=1, DMF)
Solubility Information Solubility in water: slighty soluble. Other solubilities: soluble in methanol
Formula Weight 280.3
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material D(-)-Luciferin
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-981-3

RUO – Research Use Only