accessibility menu, dialog, popup

UserName

CAS RN 2591-17-5

HOO(S)SNHSON(Z)

CAS RN 2591-17-5

IUPAC Name: (4S)-2-[(2Z)-6-oxo-2,6-dihydro-1,3-benzothiazol-2-ylidene]-1,3-thiazolidine-4-carboxylic acid
Synonyms: firefly luciferin
Molecular Weight (g/mol): 280.32
Molecular Formula: C11H8N2O3S2
InChi Key: IWJYWBVPCGUPLO-BFUDMSGGSA-N
SMILES: OC(=O)[C@H]1CS\C(N1)=C1/SC2=CC(=O)C=CC2=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)