missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Luciferin 98.0+%, TCI America™

Catalog No. p-7118261
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A503010MG 10 mg
A503050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A503010MG Supplier TCI America Supplier No. A503010MG
Only null left

Chemical Identifiers

CAS 2591-17-5
Molecular Formula C11H8N2O3S2
Molecular Weight (g/mol) 280.316
MDL Number MFCD00042929
InChI Key IWJYWBVPCGUPLO-SSDOTTSWSA-N
Synonym luciferin, d-luciferin, s-2-6-hydroxybenzo d thiazol-2-yl-4,5-dihydrothiazole-4-carboxylic acid, luciferin firefly, s-2-6-hydroxy-2-benzothiazolyl-2-thiazoline-4-carboxylic acid, 4s-2-6-oxo-1,3-benzothiazol-2-ylidene-1,3-thiazolidine-4-carboxylic acid, 4-thiazolecarboxylicacid, 4,5-dihydro-2-6-hydroxy-2-benzothiazolyl-, 4s
PubChem CID 7023066
IUPAC Name (4S)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylic acid
SMILES C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)O

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 10 mg
Formula Weight 280.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Luciferin [Chemiluminescence Reagent]
TSCA No
Recommended Storage Freezer