missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Lactic Acid 90.0+%, TCI America™
Supplier: TCI America L026625G
Specifications
| D-Lactic Acid | |
| Yellow | |
| MFCD00068311 | |
| 3265 | |
| JVTAAEKCZFNVCJ-UWTATZPHSA-N | |
| (2R)-2-hydroxypropanoic acid | |
| 61503 | |
| 90.08 | |
| Liquid |
| 10326-41-7 | |
| C3H6O3 | |
| 25 g | |
| d-lactic acid, r-2-hydroxypropanoic acid, r-lactic acid, --lactic acid, 2r-2-hydroxypropanoic acid, d-lactate, r---lactic acid, r-lactate, propanoic acid, 2-hydroxy-, 2r, d-milchsaeure | |
| CC(C(=O)O)O | |
| 90.078 | |
| CHEBI:42111 | |
| ≥90.0% (T) |
Chemical Identifiers
| 10326-41-7 | |
| 90.078 | |
| JVTAAEKCZFNVCJ-UWTATZPHSA-N | |
| 61503 | |
| (2R)-2-hydroxypropanoic acid |
| C3H6O3 | |
| MFCD00068311 | |
| d-lactic acid, r-2-hydroxypropanoic acid, r-lactic acid, --lactic acid, 2r-2-hydroxypropanoic acid, d-lactate, r---lactic acid, r-lactate, propanoic acid, 2-hydroxy-, 2r, d-milchsaeure | |
| CHEBI:42111 | |
| CC(C(=O)O)O |
Safety and Handling
EINECSNumber : (2)-1369
TSCA : Yes
Recommended Storage : Freezer