missing translation for 'onlineSavingsMsg'
Learn More

D-Lactic Acid 90.0+%, TCI America™

Catalog No. p-7131900
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L02665G 5 g
L026625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L02665G Supplier TCI America Supplier No. L02665G
Only null left

Chemical Identifiers

CAS 10326-41-7
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00068311
InChI Key JVTAAEKCZFNVCJ-UWTATZPHSA-N
Synonym d-lactic acid, r-2-hydroxypropanoic acid, r-lactic acid, --lactic acid, 2r-2-hydroxypropanoic acid, d-lactate, r---lactic acid, r-lactate, propanoic acid, 2-hydroxy-, 2r, d-milchsaeure
PubChem CID 61503
ChEBI CHEBI:42111
IUPAC Name (2R)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 90.08
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material D-Lactic Acid
EINECSNumber (2)-1369
TSCA Yes
Recommended Storage Freezer