missing translation for 'onlineSavingsMsg'
Learn More

D(-)-isovaline, 97%

Catalog No. AC388032500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC388032500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC388032500 Supplier Thermo Scientific Chemicals Supplier No. 388032500

CAS: 3059-97-0 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3059-97-0
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00800497
InChI Key GCHPUFAZSONQIV-RXMQYKEDSA-N
Synonym d-isovaline, l-isovaline, s-2-amino-2-methylbutanoic acid, l-iso-valine, unii-ebv7h5w26h, ebv7h5w26h, isovaline, l, d--isovaline, 2s-2-amino-2-methylbutanoic acid, s-2-amino-2-methylbutyric acid
PubChem CID 6971276
IUPAC Name (2S)-2-amino-2-methylbutanoic acid
SMILES CC[C@@](C)([NH3+])C([O-])=O

Specifications

Melting Point 276°C to 278°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3CH2CNH2(CH3)COOH
Quantity 250 mg
Merck Index 15, 5280
Specific Rotation -9.0° to -12.0° (20°C, 589nm) (C=1, water)
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 117.15
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material D(-)-Isovaline
Show More Show Less

RUO – Research Use Only