accessibility menu, dialog, popup

UserName

CAS RN 3059-97-0

IUPAC Name: (2R)-2-azaniumyl-2-methylbutanoate
Synonyms: (2R)-2-ammonio-2-methylbutanoate
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: GCHPUFAZSONQIV-RXMQYKEDSA-N
SMILES: CC[C@@](C)([NH3+])C([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)