missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Isoascorbic acid, 98%

CAS: 89-65-6 | C6H7NaO6 | 198.11 g/mol

Supplier:  Thermo Scientific Chemicals 227270025

 View more versions of this product

Catalog No. AC227270025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D(-)-Isoascorbic acid
89-65-6
White to Yellow
Authentic
Plastic bottle
MFCD00005378
15, 5171
− 17.25
Solubility in water: 100g/l. Other solubilities: soluble in alcohol,pyridine,moderately soluble in acetone,slightly soluble in glycerol
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
198.11
CHEBI:51438
98%
98%
167.0°C to 172.0°C
0.4% max.
97.5% min. (Iodimetry)
C6H7NaO6
2.5 kg
−17.25°(25°C c=10,H2O)
erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5
IFVCRSPJFHGFCG-HXPAKLQESA-N
(2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
54675810
176.13
Crystalline Powder or Crystals

Chemical Identifiers

89-65-6
198.11
IFVCRSPJFHGFCG-HXPAKLQESA-N
54675810
(2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
C6H7NaO6
MFCD00005378
erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5
CHEBI:51438
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-928-0

RTECSNumber : KF3015000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only