missing translation for 'onlineSavingsMsg'
Learn More

D-Iditol 98.0+%, TCI America™

Catalog No. I0724100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0724100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I0724100MG Supplier TCI America Supplier No. I0724100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent with a purity of ≥98.0% as determined by HPLC.

Chemical Identifiers

CAS 25878-23-3
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD09951922
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
PubChem CID 90540
ChEBI CHEBI:17459
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 76°C
Color White
Boiling Point 230°C
Quantity 100 mg
Formula Weight 182.17
Percent Purity ≥98.0% (HPLC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material D-Iditol