missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Histidine 99.0+%, TCI America™
$88.56 - $288.77
Chemical Identifiers
| CAS | 351-50-8 |
|---|---|
| Molecular Formula | C6H9N3O2 |
| Molecular Weight (g/mol) | 155.16 |
| MDL Number | MFCD00065963 |
| InChI Key | HNDVDQJCIGZPNO-RXMQYKEDSA-N |
| Synonym | d-histidine, h-d-his-oh, d-histidin, r-2-amino-3-1h-imidazol-4-yl propanoic acid, d-his, 2r-2-amino-3-1h-imidazol-4-yl propanoic acid, r-2-amino-3-4-imidazolyl propionic acid, 2r-2-amino-3-1h-imidazol-5-yl propanoic acid, r-alpha-amino-1h-imidazole-4-propionic acid, d-alpha-amino-beta-4-imidazolyl propionic acid |
| PubChem CID | 71083 |
| ChEBI | CHEBI:27947 |
| IUPAC Name | (2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid |
| SMILES | N[C@H](CC1=CN=CN1)C(O)=O |
Chemical Identifiers
| 351-50-8 | |
| 155.16 | |
| HNDVDQJCIGZPNO-RXMQYKEDSA-N | |
| 71083 | |
| (2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid |
| C6H9N3O2 | |
| MFCD00065963 | |
| d-histidine, h-d-his-oh, d-histidin, r-2-amino-3-1h-imidazol-4-yl propanoic acid, d-his, 2r-2-amino-3-1h-imidazol-4-yl propanoic acid, r-2-amino-3-4-imidazolyl propionic acid, 2r-2-amino-3-1h-imidazol-5-yl propanoic acid, r-alpha-amino-1h-imidazole-4-propionic acid, d-alpha-amino-beta-4-imidazolyl propionic acid | |
| CHEBI:27947 | |
| N[C@H](CC1=CN=CN1)C(O)=O |
Specifications
| 351-50-8 | |
| C6H9N3O2 | |
| d-histidine, h-d-his-oh, d-histidin, r-2-amino-3-1h-imidazol-4-yl propanoic acid, d-his, 2r-2-amino-3-1h-imidazol-4-yl propanoic acid, r-2-amino-3-4-imidazolyl propionic acid, 2r-2-amino-3-1h-imidazol-5-yl propanoic acid, r-alpha-amino-1h-imidazole-4-propionic acid, d-alpha-amino-beta-4-imidazolyl propionic acid | |
| N[C@H](CC1=CN=CN1)C(O)=O | |
| 5 g | |
| 71083 | |
| 155.16 | |
| Crystalline Powder |
| White | |
| MFCD00065963 | |
| HNDVDQJCIGZPNO-RXMQYKEDSA-N | |
| (2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid | |
| 155.16 | |
| CHEBI:27947 | |
| ≥99.0% (T,HPLC) | |
| D-Histidine |
Safety and Handling
TSCA : Yes