missing translation for 'onlineSavingsMsg'
Learn More

D-Histidine Methyl Ester Dihydrochloride 97.0+%, TCI America™

Catalog No. p-7130923
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12131G 1 g
H12135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12131G Supplier TCI America Supplier No. H12131G

Chemical Identifiers

CAS 4467-54-3
Molecular Formula C7H13Cl2N3O2
Molecular Weight (g/mol) 242.1
MDL Number MFCD00066137
InChI Key DWAYENIPKPKKMV-QYCVXMPOSA-N
Synonym d-histidine methyl ester dihydrochloride, r-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, h-d-his-ome 2hcl, h-d-his-ome.2hcl, d-histidine methylester dihydrochloride, c7h11n3o2.2hcl, methyl 2r-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, d-histidine methyl ester di hydrochloride, d-his-ome.2hcl, d-histidinemethylesterdihydrochloride
PubChem CID 12658398
IUPAC Name methyl (2R)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
SMILES COC(=O)C(CC1=CN=CN1)N.Cl.Cl

Specifications

Melting Point 197°C
Color White
Quantity 1 g
Formula Weight 242.10
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material D-Histidine Methyl Ester Dihydrochloride