accessibility menu, dialog, popup

UserName

H3COONHNNH2HClHCl

CAS RN 4467-54-3

IUPAC Name: methyl 2-amino-3-(1H-imidazol-5-yl)propanoate dihydrochloride
Synonyms: methyl 2-amino-3-(3H-imidazol-4-yl)propanoate dihydrochloride
Molecular Weight (g/mol): 242.1
Molecular Formula: C7H13Cl2N3O2
InChi Key: DWAYENIPKPKKMV-UHFFFAOYNA-N
SMILES: Cl.Cl.COC(=O)C(N)CC1=CN=CN1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)