missing translation for 'onlineSavingsMsg'
Learn More

D-Histidine methyl ester dihydrochloride, 95%

CAS: 4467-54-3 | C7H13Cl2N3O2 | 242.1 g/mol

$56.85 - $762.62

Chemical Identifiers

CAS 4467-54-3
Molecular Formula C7H13Cl2N3O2
Molecular Weight (g/mol) 242.1
MDL Number MFCD00066137
InChI Key DWAYENIPKPKKMV-QYCVXMPOSA-N
Synonym d-histidine methyl ester dihydrochloride, r-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, h-d-his-ome 2hcl, h-d-his-ome.2hcl, d-histidine methylester dihydrochloride, c7h11n3o2.2hcl, methyl 2r-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, d-histidine methyl ester di hydrochloride, d-his-ome.2hcl, d-histidinemethylesterdihydrochloride
PubChem CID 12658398
IUPAC Name methyl (2R)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
SMILES COC(=O)C(CC1=CN=CN1)N.Cl.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6509603
View Documents
Thermo Scientific Chemicals
H6509603
1 g
Each for $56.85
Only null left
 
AAH6509606
View Documents
Thermo Scientific Chemicals
H6509606
5 g
Each for $178.30
Only null left
 
AAH6509614
View Documents
Thermo Scientific Chemicals
H6509614
25 g
Each for $762.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4467-54-3
242.1
DWAYENIPKPKKMV-QYCVXMPOSA-N
12658398
COC(=O)C(CC1=CN=CN1)N.Cl.Cl
C7H13Cl2N3O2
MFCD00066137
d-histidine methyl ester dihydrochloride, r-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, h-d-his-ome 2hcl, h-d-his-ome.2hcl, d-histidine methylester dihydrochloride, c7h11n3o2.2hcl, methyl 2r-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, d-histidine methyl ester di hydrochloride, d-his-ome.2hcl, d-histidinemethylesterdihydrochloride
methyl (2R)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride

Specifications

4467-54-3
C7H13Cl2N3O2
6491805
DWAYENIPKPKKMV-QYCVXMPOSA-N
methyl (2R)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
242.1
242.1
95%
D-Histidine methyl ester dihydrochloride
White
MFCD00066137
d-histidine methyl ester dihydrochloride, r-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, h-d-his-ome 2hcl, h-d-his-ome.2hcl, d-histidine methylester dihydrochloride, c7h11n3o2.2hcl, methyl 2r-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, d-histidine methyl ester di hydrochloride, d-his-ome.2hcl, d-histidinemethylesterdihydrochloride
COC(=O)C(CC1=CN=CN1)N.Cl.Cl
1 g
12658398
Hygroscopic
Solid
Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only