missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamic acid dimethyl ester hydrochloride, 98%

CAS: 27025-25-8 | C7H14ClNO4 | 211.642 g/mol

$46.42 - $540.18

Chemical Identifiers

CAS 27025-25-8
Molecular Formula C7H14ClNO4
Molecular Weight (g/mol) 211.642
MDL Number MFCD00235872
InChI Key MFUPLHQOVIUESQ-NUBCRITNSA-N
Synonym dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride
PubChem CID 12917567
IUPAC Name dimethyl (2R)-2-aminopentanedioate;hydrochloride
SMILES COC(=O)CCC(C(=O)OC)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6213203
View Documents
Thermo Scientific Chemicals
H6213203
1 g
Each for $46.42
Only null left
 
AAH6213206
View Documents
Thermo Scientific Chemicals
H6213206
5 g
Each for $116.15
Only null left
 
AAH6213214
View Documents
Thermo Scientific Chemicals
H6213214
25 g
Each for $540.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

27025-25-8
211.642
MFUPLHQOVIUESQ-NUBCRITNSA-N
12917567
COC(=O)CCC(C(=O)OC)N.Cl
C7H14ClNO4
MFCD00235872
dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride
dimethyl (2R)-2-aminopentanedioate;hydrochloride

Specifications

91°C
C7H14ClNO4
dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride
COC(=O)CCC(C(=O)OC)N.Cl
1 g
12917567
211.64
98%
27025-25-8
MFCD00235872
MFUPLHQOVIUESQ-NUBCRITNSA-N
dimethyl (2R)-2-aminopentanedioate;hydrochloride
211.642
−26° (c=5 in Water)
Hygroscopic
D-Glutamic acid dimethyl ester hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only