missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glutamic acid dimethyl ester hydrochloride, 98%
CAS: 27025-25-8 | C7H14ClNO4 | 211.642 g/mol
Supplier: Thermo Scientific Chemicals H6213206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 91°C | |
| 27025-25-8 | |
| MFCD00235872 | |
| MFUPLHQOVIUESQ-NUBCRITNSA-N | |
| dimethyl (2R)-2-aminopentanedioate;hydrochloride | |
| 211.642 | |
| −26° (c=5 in Water) | |
| Hygroscopic |
| D-Glutamic acid dimethyl ester hydrochloride | |
| C7H14ClNO4 | |
| dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride | |
| COC(=O)CCC(C(=O)OC)N.Cl | |
| 5 g | |
| 12917567 | |
| 211.64 | |
| 98% |
Chemical Identifiers
| 27025-25-8 | |
| 211.642 | |
| MFUPLHQOVIUESQ-NUBCRITNSA-N | |
| 12917567 | |
| COC(=O)CCC(C(=O)OC)N.Cl |
| C7H14ClNO4 | |
| MFCD00235872 | |
| dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride | |
| dimethyl (2R)-2-aminopentanedioate;hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only