Learn More
D-Glucurono-6,3-lactone, 99%
CAS: 32449-92-6 | C6H8O6 | 176.124 g/mol
Supplier: Thermo Scientific Chemicals A1586114
Description
D-Glucurono-6,3-lactone is a glucuronic acid derivative studied for its effectiveness against canine hepatitis. It is utilized in the studies such as starting regent in the synthesis of 2,3,4,-tris(tert.-butyldimethysilyl) glucuronic acid trichloroethylester, required for the preparation of 1-O-acyl glucuronide of the anti-inflammatory drug ML-3000, synthesis of optically active glucopyranoses, synthesis of long-chain alkyl glucofuranosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Glucurono-6,3-lactone is a glucuronic acid derivative studied for its effectiveness against canine hepatitis. It is utilized in the studies such as starting regent in the synthesis of 2,3,4,-tris(tert.-butyldimethysilyl) glucuronic acid trichloroethylester, required for the preparation of 1-O-acyl glucuronide of the anti-inflammatory drug ML-3000, synthesis of optically active glucopyranoses, synthesis of long-chain alkyl glucofuranosides.
Solubility
Soluble in Water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong acids and strong oxidizing agents.
Specifications
| D-Glucurono-6,3-lactone | |
| 174°C (decomposition) | |
| MFCD00135622 | |
| 83595 | |
| d-glucuro-3,6-lactone | |
| UYUXSRADSPPKRZ-KKQCNMDGSA-N | |
| C(=O)C(C1C(C(C(=O)O1)O)O)O | |
| 176.124 | |
| 176.12 |
| 32449-92-6 | |
| C6H8O6 | |
| 25 g | |
| 14,4467 | |
| Soluble in Water. | |
| +19° (c=8 in Water) | |
| (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde | |
| 91634076 | |
| 99% |
Chemical Identifiers
| 32449-92-6 | |
| 176.124 | |
| UYUXSRADSPPKRZ-KKQCNMDGSA-N | |
| 91634076 | |
| C(=O)C(C1C(C(C(=O)O1)O)O)O |
| C6H8O6 | |
| MFCD00135622 | |
| d-glucuro-3,6-lactone | |
| (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde |
Safety and Handling
EINECSNumber : 251-053-3
RTECSNumber : LZ8930000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only