missing translation for 'onlineSavingsMsg'
Learn More

D-Glucurono-6,3-lactone, 99%

Catalog No. p-7022889
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1586114 25 g
AAA1586122 100 g
AAA1586136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1586114 Supplier Thermo Scientific Chemicals Supplier No. A1586114
Only null left

CAS: 32449-92-6 | C6H8O6 | 176.124 g/mol

D-Glucurono-6,3-lactone is a glucuronic acid derivative studied for its effectiveness against canine hepatitis. It is utilized in the studies such as starting regent in the synthesis of 2,3,4,-tris(tert.-butyldimethysilyl) glucuronic acid trichloroethylester, required for the preparation of 1-O-acyl glucuronide of the anti-inflammatory drug ML-3000, synthesis of optically active glucopyranoses, synthesis of long-chain alkyl glucofuranosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucurono-6,3-lactone is a glucuronic acid derivative studied for its effectiveness against canine hepatitis. It is utilized in the studies such as starting regent in the synthesis of 2,3,4,-tris(tert.-butyldimethysilyl) glucuronic acid trichloroethylester, required for the preparation of 1-O-acyl glucuronide of the anti-inflammatory drug ML-3000, synthesis of optically active glucopyranoses, synthesis of long-chain alkyl glucofuranosides.

Solubility
Soluble in Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong acids and strong oxidizing agents.

Chemical Identifiers

CAS 32449-92-6
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.124
MDL Number MFCD00135622
InChI Key UYUXSRADSPPKRZ-KKQCNMDGSA-N
Synonym d-glucuro-3,6-lactone
PubChem CID 91634076
IUPAC Name (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
SMILES C(=O)C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 174°C (decomposition)
Quantity 25 g
Beilstein 83595
Merck Index 14,4467
Solubility Information Soluble in Water.
Optical Rotation +19° (c=8 in Water)
Formula Weight 176.12
Percent Purity 99%
Chemical Name or Material D-Glucurono-6,3-lactone
EINECSNumber 251-053-3
RTECSNumber LZ8930000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only