missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Glucurono-3,6-lactone, 99+%
CAS: 32449-92-6 | C6H8O6 | 176.12 g/mol
Supplier: Thermo Scientific Chemicals 225921000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| D(+)-Glucurono-3, 6-lactone | |
| 32449-92-6 | |
| White | |
| Authentic | |
| Plastic bottle | |
| MFCD00135622 | |
| 15, 4503 | |
| + 19.00 | |
| (5% in water) Clear colorless to light yellow | |
| C(=O)C(C1C(C(C(=O)O1)O)O)O | |
| 176.12 | |
| 176.12 | |
| Crystalline Powder or Crystals |
| 99+% | |
| 170.0°C to 176.0°C | |
| 0.5% max. (80°C) | |
| 99+% | |
| C6H8O6 | |
| 100 g | |
| +19.00 (20.00°C c=10,H2O) | |
| d-glucuro-3,6-lactone | |
| UYUXSRADSPPKRZ-KKQCNMDGSA-N | |
| (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde | |
| 91634076 | |
| 99+% |
Chemical Identifiers
| 32449-92-6 | |
| UYUXSRADSPPKRZ-KKQCNMDGSA-N | |
| 91634076 | |
| C(=O)C(C1C(C(C(=O)O1)O)O)O |
| MFCD00135622 | |
| d-glucuro-3,6-lactone | |
| (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde |
Safety and Handling
EINECSNumber : 251-053-3
RTECSNumber : LZ8930000
TSCA : TSCA
RUO – Research Use Only