missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucurono-3,6-lactone, 99+%

Catalog No. p-138081
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225920250 25 g Plastic bottle
AC225921000 100 g Plastic bottle
AC225925000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC225920250 Supplier Thermo Scientific Chemicals Supplier No. 225920250
Only null left

CAS: 32449-92-6 | C6H8O6 | 176.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32449-92-6
MDL Number MFCD00135622
InChI Key UYUXSRADSPPKRZ-KKQCNMDGSA-N
Synonym d-glucuro-3,6-lactone
PubChem CID 91634076
IUPAC Name (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
SMILES C(=O)C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 170.0°C to 176.0°C
Color White
Loss on Drying 0.5% max. (80°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Molecular Formula C6H8O6
Quantity 25 g
Merck Index 15, 4503
Specific Rotation Condition +19.00 (20.00°C c=10,H2O)
Specific Rotation + 19.00
Solubility Information (5% in water) Clear colorless to light yellow
Molecular Weight (g/mol) 176.12
Formula Weight 176.12
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(+)-Glucurono-3, 6-lactone, 99+%
Show More Show Less
EINECSNumber 251-053-3
RTECSNumber LZ8930000
TSCA TSCA

RUO – Research Use Only